Would you like to log in again?
EMbed 812, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14900 14901
100503-878EA 242.63 USD InStock
100503-878 100503-876
EMbed 812, Electron Microscopy Sciences
Stains Microscopy and staining reagents
EMbed 812 has the same characteristics as Epon 812, physically, and provides the same results in preservation, handling, curing and sectioning.
  • Specific Gravity: 1.08-1.10
EMbed 812 may cause etching on selected plastics. All formulas for mixing and using Epon 812 can be applied to EMbed 812, e.g. Luft's procedure, Mollenhauer's and EPON-ARALDITE mixtures.
Ordering information: Certificates of analysis are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty