Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluidine blue - Basic fuchsin solution Epoxy Tissue Stain

Supplier: Electron Microscopy Sciences
Danger

14950
100503-882EA 53.66 USD InStock
100503-882
Toluidine blue - Basic fuchsin solution Epoxy Tissue Stain
Toluidine blue - Basic fuchsin solution
A toluidine blue and basic fuchsin preparation; ready to use. For staining semi-thin sections of epoxy embedded tissues.

Spurlock, B.O., (1966) et al., Am. J. of Clin. Path.


Boiling Pt: 78 °C
Flash Pt: 79 °C
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty