Would you like to log in again?
Bouin's Fixative, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15990
100503-962EA 77.64 USD InStock
100503-962
Bouin's Fixative, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Bouin's Fixative is used in light microscopy for the preparation of delicate structure
Bouin's Fixative is used in light microscopy for the preparation of delicate structure. Formulated with Picric Acid saturated aqueous solution 75%, Formalin (40% aqueous formaldehyde) 25% and Glacial Acetic acid 5% in de-ionized water
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty