Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Methyl methacrylate

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Methacrylic acid methyl ester

18800
100504-110EA 15.38 USD InStock
100504-110
Methyl methacrylate
Methyl methacrylate (monomer)
Inhibitor (Hydroquinone): 0.0025% (25 ppm).


Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 101 °C (214 °F)
Melting Pt: –48 °C (–54 °F)
Density: 0.94 g/cm³ (7.844 lbs/gal.)
Flash Pt: 10 °C (50 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008587
CAS Number: 80-62-6
UN: 1247
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/55/28984855.jpg
Order Now
Specification Test Results
Specific gravity at 20 °C 0.943

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty