Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Paraformaldehyde, granular, EM Grade, purified

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Polyoxymethylene

19210 19208
100504-160EA 40.3 USD InStock
100504-160 100504-162
Paraformaldehyde, granular, EM Grade, purified
Paraformaldehyde
Paraformaldehyde is the condensation product of methylene glycol, a substance not so far isolated in its pure form; it is a white solid with a formaldehyde content between 78 and 98% and is supplied either as a Prill or Granular.

All forms contain more than 95.5% paraformaldehyde content. The Prill and Granular forms are small, free-flowing, dust-free beads, the most desired by EM users.
A fast-penetrating EM fixative used in combination with Glutaraldehyde, Acrolein, and Osmium Tetroxide.
Karnovsky, M.J., J. Cell Biol., 27, 137A (1965). Hayat, M.A. Principles & Tech., of EM (Univ. Park Press), 68 (1981).


Formula: (CH₂O)n
Melting Pt: 120 °C (248 °F)
MDL Number: MFCD00133991
CAS Number: 30525-89-4
UN: 2213
ADR: 4.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/71/28984871.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty