Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Potassium hydroxide, pellets ACS

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Potassium hydrateCaustic potash

20155
100504-232EA 32.58 USD InStock
100504-232
Potassium hydroxide, pellets ACS
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1327 °C (2421 °F)
Melting Pt: 360 °C (680 °F)
Density: 2.04 g/cm³ (17.024 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II
Order Now
Specification Test Results
Assay 85.0%
Water contains 10-15%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty