Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium iodide, granules ACS

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Dipotassium diiodideKali iodide

20158
100504-234EA 80.26 USD InStock
100504-234
Potassium iodide, granules ACS
Potassium iodide
Refer to bismuth carbonate for references on use.


Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (2426 °F)
Melting Pt: 681 °C (1258 °F)
Density: 3.13 g/cm³ (26.12 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/84/28984884.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty