Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥97.0%, pellets ACS

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

21160 21162
100504-282EA 16.24 USD InStock
100504-282 100504-284
Sodium hydroxide ≥97.0%, pellets ACS
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (2534 °F)
Melting Pt: 318 °C (604 °F)
Density: 2.13 g/cm³ (17.775 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Ammonium hydroxide 0.02% ppt
Assay 97.0%
Chloride 0.005%
Heavy metals 0.002%
Iron 0.001%
Mercury 0.1 ppm
Nickel 0.001%
Nitrogen compounds 0.001%
Phosphate 0.001%
Potassium 0.02%
Sodium carbonate 1%
Sulfate 0.003%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty