Would you like to log in again?
Apiezon Grease; Type T is, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60704
100504-356EA 381.47 USD InStock
100504-356
Apiezon Grease; Type T is, Electron Microscopy Sciences
Lubricants
Type T is designed for lubricating glass taps and for general purposes. Has a melting point of 125°C and has an estimated vapor pressure @ 20°C of 5x10-9 torr. Although type T is useful at higher temperatures, it can also be used at ambient temperatures. Temperature ranges 0-120°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty