Would you like to log in again?
Santovac 5, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60730 60732
100504-362EA 596.79 USD InStock
100504-362 100504-364
Santovac 5, Electron Microscopy Sciences
Lubricants
A Polymethyl Ether, designed for an ultra high vacuum range: 10-10 torr. Ultra low vapor pressure results in low ultimate pressures; there is no need for a cold trap and it is suitable for diffusion pumps. Santovac 5 may discolor with use but it will maintain its physical properties.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty