Would you like to log in again?
Microtome Oil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60744
100504-370EA 207.64 USD InStock
100504-370
Microtome Oil, Electron Microscopy Sciences
Lubricants
A chemically pure, high-grade lubricant used in precision instruments. Should be used periodically to lubricate cross-rollerbearings for microtomes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty