Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 2 N in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

21170 21170-01
100504-964EA 31.43 USD InStock
100504-964 100504-376
Sodium hydroxide 2 N in aqueous solution
Sodium hydroxide
2N Standardized and Carbonate-free. This solution is prepared from ACS Reagent grade sodium hydroxide, and is standardized colormetrically against potassium biphthalate primary reference material.
Normal Concentration 1.98-2.02


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty