Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.01N in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Muriatic acid

26357-03
100504-598EA 33.97 USD InStock
100504-598
Hydrochloric acid 0.01N in aqueous solution
Hydrochloric acid
Picrosirius Red Staining of Cardiac Muscle Following Phosphomolybdic Acid Treatment.
For connective tissue stains – Van Gieson stains and variants. Septa is stained red.

This method eliminates cytoplasmic staining, revealing thin septa and collegen fibers clearly.
Fixation: Bouins' Fixative
Reagents
26357-01 Phosphomolybdic Acid, 0.2% Aqueous
26357-02 Sirius Red (Direct Red 80), 0.1% in Saturated Picric Acid
26357-03 Hydrochloric Acid, 0.01N

References: Dolber, Paul C. and Madison S. Spach. Picrosirus Red Staining of Cardiac Muscle Following Phosphomolybdic Acid Treatment, Stain Technology, Vol. 62 No 1 (1987) 23 – 26.

Certifications: Certificates of analysis are available from Electron Microscopy Sciences. Please contact us or visit our website to search by lot or product number.


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/35/28984835.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty