Would you like to log in again?
Modified Movat’s Stain, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26385-07 26385-06 26385-09 26385-01 26385-05 26385-03 26385-04 26385-10 26385-08 26385-02
100504-610EA 55.72 USD InStock
100504-610 100504-622 100504-612 100504-624 100504-614 100504-616 100504-606 100504-618 100504-608 100504-620
Modified Movat’s Stain, Electron Microscopy Sciences
Stains Histology Stains
Modified Movat’s Stain, is used for connective tissue stains – Elastic tissue stains
Stain results - Nuclei: Black, Cytoplasm: Red, Elastic Fibers: Purple to Black, Collagen and Bone: Yellow, Mucopolysaccharides: Blue-Green, Muscle: Red.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty