Would you like to log in again?
Xylene Substitute, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
23410-05 23410-04 23410-01
100504-382EA 200.45 USD InStock
100504-382 100504-384 100504-780
Xylene Substitute, Electron Microscopy Sciences
Stains Microscopy and staining reagents
This EMS Xylene Substitute solution is specially designed for use in processing and staining histological/cytological specimens. It is essentially odorless with low toxicity, less irritating to the skin than xylene- and d-limonene-based clearants. It can be substituted for xylene and can be used with almost all tissue processors and automated slide stainers.
  • Minimal tissue shrinkage
  • Dries slowly with no residue
  • Less toxic than xylene
  • Soluble with alcohol, paraffin embedding media, and mounting media
  • Biodegradable
Xylene substitute for histology and cytology is a safer alternative to xylene. Can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. It can be used in all procedures that require xylene.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty