Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Osmic acid ≥99.95%, crystalline, highest purity

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Osmic acid anhydrideOsmic acidOsmium (VIII) oxide

19130 19112 19110 19114 19100 19105 19103 19132 19134 19102 19115 19131 19116 19120 19104
100504-130EA 228.9 USD InStock
100504-130 100504-132 100504-134 100504-136 100504-820 100504-810 100504-128 100504-812 100504-802 100504-814 100504-804 100504-816 100504-806 100504-818 100504-808
Osmic acid ≥99.95%, crystalline, highest purity
Osmium tetraoxide
Small size (2 ml), light-gauge, pre-scored amber glass ampoules containing OsO₄ crystal, are pre-cleaned and heat sealed in plastic bags to assure a contaminant-free solution.

Ampoules are also free of pressure-sensitive labels and, therefore, do not require cleaning prior to solution preparation.

Solubility 25.0% in CCl4
Solubility in water: 5.07% in Cold Water; 6.5% in Hot Water; Moderately sol. (7 g/100 mL)


Formula: O₄Os
MW: 254.23 g/mol
Boiling Pt: 130 °C
Melting Pt: 40.1 °C
Density: 4.906 g/cm³ (22°C)
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 2471
ADR: 6.1,I
Order Now
Specification Test Results
Vapor Pressure at 25 °C 9.8 mmHg
Osmium Purity 99.95%
Non-Volatile Matter 0.02%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty