Would you like to log in again?
Apiezon Grease; Type H, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60703
100504-876EA 392.53 USD InStock
100504-876
Apiezon Grease; Type H, Electron Microscopy Sciences
Lubricants
Type H grease is designed for high temperatures (-10 to +240°C) in a wide range of applications in both science and industry. With good "friction", grease H is ideal for use with laboratory glassware, but combined with properties of high thermal conductivity, it is the perfect choice for the electronics and space industries where heat sink media require adhesion. Grease H is a filled hydrocarbon which exhibits excellent thermal conductivity, and strong absorption properties. The capability to absorb greasy or chemical impurities on metal and glass surfaces is a value "tool" required by electronic industries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty