Would you like to log in again?
Oil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60720 60724 60722
100504-974EA 339.14 USD InStock
100504-974 100504-878 100504-360
Oil, Electron Microscopy Sciences
Lubricants
Invoil Oil BW
  • Consists of esters or specially selected hydrocarbon fractions
  • High purity, very low pressure, very stable
  • Pressures lower than 0.013 – 0.009 bar obtainable
  • No need for cold trap
  • Good pumping speed
  • No corrosive deposits or vapors
Invoil Oil C
Obtains the highest vacuum pressure (0.013 – 0.009 bar) of all of the oils without the need of a cold trap.
Invoil Oil BW: A water-white oil with a higher pumping speed than oil B, although all other characteristics are similiar to B. Because it has an extremely high purity it is suitable for use in the evacuation of mass spectrometers.
Invoil Oil C: Obtains the highest vacuum pressure (0.013 – 0.009 bar) of all of the oils without the need of a cold trap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty