Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.5% in 70% Alcohol

Supplier: Electron Microscopy Sciences
Synonyms: Muriatic acid

26071-04 26071-01
100504-882EA 268.88 USD InStock
100504-882 100504-454
Hydrochloric acid 0.5% in 70% Alcohol
Hydrochloric acid
An acidified alcohol solution for the differentiation of regressive stains.


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty