Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ACS

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Methane, trichloro-TrichloroformTrichloromethane

12540 12541
100504-934EA 141.19 USD InStock
100504-934 100503-758
Chloroform ACS
Chloroform
CHCl3 FW 119.38 CAS 67-66-3 A.C.S.


Formula: CHCl₃
MW: 119.38 g/mol
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/98/28984798.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty