Would you like to log in again?
Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26123-11 26123-03 26123-07 26123-02 26123-09 26123-06 26123-01 26123-08 26123-01 26123-05 26123-10 26123-04
100504-984EA 33.16 USD InStock
100504-984 100504-544 100504-986 100504-546 100504-988 100504-548 CA11028-246 100504-538 100505-040 100504-550 100504-540 100504-542
Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Stains Histology Stains
A Combined Hematoxylin and Eosin/Methenamine Silver Stain is used for the Histological Diagnosis of Fungi in Tissue Sections.
  • For bacterial, fungal stains
Stain results - Organism: Blue-Black Background: Rose.
Lamps, Laura W. et al A Combined Hematoxylin and Eosin/Methenamin Silver Stain for theHistological Diagnosis of Fungi In Tissue Sections., J. of Histotechnology 23:4 p.341 – 345, 2000.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty