Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid solution 0.1 N

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Muriatic acidHydrochloric acid solution

16760
100505-060EA 10.98 USD InStock
100505-060
Hydrochloric acid solution 0.1 N
Hydrochloric acid
Used to adjust the pH of buffers and fixative solutions.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 1.003 g/cm³ (8.37 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/35/28984835.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty