Would you like to log in again?
Taylor’s Method for Gram Positive and Gram Negative Bacteria, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26108-06 26108-01 26108-10 26108-07 26108-05 26108-03 26108-08 26108-02 26108-04 26108-09 26108-11
100505-068EA 56.72 USD InStock
100505-068 100504-512 100505-066 100504-514 100505-064 100504-516 100504-518 100504-508 100505-072 100505-070 100504-510
Taylor’s Method for Gram Positive and Gram Negative Bacteria, Electron Microscopy Sciences
Stains Microbiology Stains
Taylor’s Method is used for gram positive and gram negative bacteria
Stain results - Gram-Positive Organism: Blue to Blue-Black, Gram Negative Organism: Bright Red, Nuclei: Brownish Red, Erythrocytes: Red to Yellow-Green.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty