Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2F1]

Supplier: Tonbo Biosciences
35-5893-U025 35-5893-U500 35-5893-U100
10051-012EA 163 USD InStock
10051-012 10051-010 10141-160
Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2F1]
Antibodies
Type: Primary
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty