Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]

Supplier: Tonbo Biosciences
40-0161-U100 40-0161-M001 40-0161-U500
10051-078EA 348 USD InStock
10051-078 10051-080 10051-082
Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Antibodies
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). As these are receptors for the Fc portion of mouse IgG, they may also bind laboratory antibody preparations and products used in a variety of cell analysis protocols such as flow cytometry, immunohistochemistry and functional cell assays. The 2.4G2 antibody is therefore widely used as a pre-treatment reagent to block binding of specific antibodies of interest, e.g. fluorescently conjugated antibodies, to Fc receptors via their Fc domains and contributing to “non-specific” staining.

Type: Primary
Antigen: CD16 + CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty