Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]

Supplier: Tonbo Biosciences
40-0281-M001 40-0281-U100 40-0281-U500
10051-090EA 114 USD InStock
10051-090 10051-088 10051-092
Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]
Antibodies
The 37.51 antibody reacts with mouse CD28, a 45 kDa glycoprotein which acts as a co-stimulatory receptor in support of the T cell receptor (TCR). CD28 exists as a homodimer with specificity for two known ligands, known as B7-1 (CD80) and B7-2 (CD86), expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. Other members of the CD28 family of co-stimulatory receptors include CTLA-4 (CD152), PD-1 (CD279), ICOS and BTLA.

The 37.51 may be used as a phenotypic marker for CD28, which is expressed on all CD4+ T cells and CD8+ T cells, and on NK cells in mouse. In addition, the 37.51 antibody is widely used to activate the CD28 receptor in vitro and in vivo. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 37.51
Conjugation:
Epitope:
Host: Golden Syrian Hamster
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty