Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFNG Rat Monoclonal Antibody [clone: XMG1.2]

Supplier: Tonbo Biosciences
40-7311-U500 40-7311-U100
10051-136EA 115 USD InStock
10051-136 10051-138
Anti-IFNG Rat Monoclonal Antibody [clone: XMG1.2]
Antibodies
The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20 kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells, which allow the cytokine to modulate innate immunity, when secreted by NK and NKT cells, and to function in supporting adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).

The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein by flow cytometry. Other formats can be used for quantitative analysis of the secreted protein by ELISA, when paired with an appropriated secondary antibody. This clone has been reported for neutralization of the functional activity of IFN-g in a variety of assays (use format suitable for functional assays).

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: XMG1.2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty