Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-6.7]

Supplier: Tonbo Biosciences
50-0081-U500 50-0081-U100 50-0081-U025
10051-184EA 223 USD InStock
10051-184 10051-180 10051-182
Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-6.7]
Antibodies
The 53-6.7 antibody reacts with the 32-34 kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.

The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty