Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: PC61.5]

Supplier: Tonbo Biosciences
50-0251-U025 50-0251-U100 50-0251-U500
10051-226EA 43.9 USD InStock
10051-226 10002-294 10051-228
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: PC61.5]
Antibodies
The PC61.5 antibody is specific for mouse CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Expression of CD25 varies during developmental stages of T and B cells, is induced on activated mature T and B cells, and is present on subsets of dendritic cells. CD25 signaling as part of the IL-2 receptor complex triggers T cell activation and proliferation, as well as modulating the differentiation and function of Th17 cells, T regulatory (Treg) cells, and dendritic cells.

The PC61.5 antibody is used as a marker for T cells, B cells and dendritic cell subsets. Expression of CD25, CD4 and the transcription factor Foxp3 is regarded as a phenotypic signature for Treg cells. As such, this antibody is widely used to distinguish Treg cells from naïve or conventional T cells which are CD25-. This clone has also been reported for depletion of Treg cells in vivo (use format suitable for functional assays).

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: PC61.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty