Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTLA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BNI3]

Supplier: Tonbo Biosciences
50-1529-T100 50-1529-T025
10051-314EA 73.5 USD InStock
10051-314 10051-316
Anti-CTLA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BNI3]
Antibodies
The BNI3 antibody is specific for human CD152, commonly known as CTLA-4, a 33-37 kDa protein expressed as a homodimer on the surface of activated T and B cells, and on thymocytes. CTLA-4 is structurally similar, yet functionally disparate, to the T cell co-stimulatory molecule CD28. Both CTLA-4 and CD28 interact with the co-stimulatory molecules CD80 (B7-1) and CD86 (B7-2) on antigen-presenting cells, with CTLA-4 displaying a higher avidity than CD28. While CD28 typically delivers a potent co-stimulatory signal in support of T cell activation, CTLA-4 appears to act as a negative regulator of T cell activation and may contribute to the suppressor function of Treg cells.

CTLA-4 proteins may be initially sequestered within Golgi vesicles, from which they can be transferred to and from the cell surface, a mechanism by which Treg cells can selectively impart suppressive functions. The BNI3 antibody may be used for flow cytometric analysis of intracellular or surface CTLA-4 expression, and is also widely used for neutralization of CTLA-4 when expressed at the cell surface. The BNI3 antibody is reported to be cross-reactive with Baboon, Cynomolgus and Rhesus CTLA-4. Please choose the appropriate format for each application.

Type: Primary
Antigen: CTLA4
Clonality: Monoclonal
Clone: BNI3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty