Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF18 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DTA-1]

Supplier: Tonbo Biosciences
50-5874-U025 50-5874-U100
10051-346EA 75.7 USD InStock
10051-346 10051-348
Anti-TNFRSF18 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DTA-1]
Antibodies
The DTA-1 antibody reacts with mouse CD357, also known as GITR or AITR (in humans), a 66-70 kDa member of the Tumor Necrosis Factor superfamily (TNFRSF18). GITR is primarily found on T cells, and its function may vary depending on the T cell type where it is expressed. GITR is upregulated on activated T cells where it provides co-stimulation, yet GITR may promote the inhibition of CD4+ CD25+ Treg cells, where it is expressed at high levels. GITR ligand (GITRL) is found on B cells, macrophages, dendritic and endothelial cells, and is implicated in regulating both innate and adaptive immune responses.

The DTA-1 antibody may be used for analysis of GITR expression on T cells, and is also commonly used in vitro as an agonistic antibody to induce GITR signaling in various assays.

Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: DTA-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty