Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LPAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]

Supplier: Tonbo Biosciences
50-5887-U100 50-5887-U025
10051-352EA 150 USD InStock
10051-352 10051-350
Anti-LPAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]
Antibodies
The DATK32 antibody is specific for a combinatorial epitope of the mouse integrin alpha 4/beta 7 (LPAM-1) heterodimer. LPAM-1 is expressed on most mature lymphocytes and sub-populations of thymocytes and bone marrow cells. It is involved in lymphocyte adhesion and migration and is a receptor for several extracellular matrix proteins including VCAM-1, fibronectin, and MAdCAM-1.

DATK32 is reported to induce LPAM-1 dependent lymphocyte aggregation and block adhesion to VCAM-1, fibronectin, and MAdCAM-1.

Type: Primary
Antigen: LPAM-1
Clonality: Monoclonal
Clone: DATK32
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty