Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: JES6-5H4]

Supplier: Tonbo Biosciences
50-7021-U100 50-7021-U025
10051-374EA 58.1 USD InStock
10051-374 10051-376
Anti-IL2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: JES6-5H4]
Antibodies
The JES6-5H4 antibody binds to mouse Interleukin-2 (IL-2), a 17 kDa cytokine that is secreted by activated T cells. This cytokine acts as an autocrine stimulatory factor for T cell proliferation, differentiation and survival, as well as being an effective activator of B cells and inducer of NK cell cytotoxic functions. IL-2 plays a key modulatory role in the differentiation of T cells toward either Th17 or T regulatory (Treg) cell types and is important for Treg cell function. The IL-2 receptor (IL-2R) is a complex consisting of an IL-2R alpha chain (CD25), along with the IL-2R beta chain (CD122) and the common gamma chain (CD132). IL-2 may also bind with low affinity to the T cell surface protein CD25 alone, although this interaction does not induce cell signaling.

The JES6-5H4 antibody is extensively used for intracellular detection of IL-2 by flow cytometry, for detection of soluble cytokine in ELISA, and as a neutralizing reagent in vitro and in vivo.

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty