Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD44 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: IM7]

Supplier: Tonbo Biosciences
65-0441-U100 65-0441-U025
10051-594EA 170 USD InStock
10051-594 10051-592
Anti-CD44 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: IM7]
Antibodies
The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or “standard” form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.

The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty