Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]

Supplier: Tonbo Biosciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

70-0161-U500 70-0161-U100 70-0161-M001
10051-664EA 139 USD InStock
10051-664 10051-660 10051-662
Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Antibodies
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). As these are receptors for the Fc portion of mouse IgG, they may also bind laboratory antibody preparations and products used in a variety of cell analysis protocols such as flow cytometry, immunohistochemistry and functional cell assays. The 2.4G2 antibody is therefore widely used as a pre-treatment reagent to block binding of specific antibodies of interest, e.g. fluorescently conjugated antibodies, to Fc receptors via their Fc domains and contributing to “non-specific” staining.

Type: Primary
Antigen: CD16 + CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty