Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KIT Rat Monoclonal Antibody [clone: ACK2]

Supplier: Tonbo Biosciences
70-1172-U100
10051-706EA 97.5 USD InStock
10051-706
Anti-KIT Rat Monoclonal Antibody [clone: ACK2]
Antibodies
The ACK2 antibody is specific for CD117, also called c-Kit, a 145 kDa cytokine receptor important in the development of hematopoietic stem cells, in oogenesis, and for functional activity of immune cells such as NK and mast cells. c-Kit binds to a ligand known as stem cell factor (SCF), or alternatively as mast cell growth factor. Ligand binding promotes the activation (dimerization) and subsequent tyrosine kinase activity of the c-Kit receptor and triggers key survival, expansion and maturation signals during hematopoietic progenitor cell development. Conversely, shedding of extracellular domain of c-Kit receptor is reported to induce inactivation or apoptosis within these cells. The survival signaling activity of c-Kit confers a proto-oncogenic attribute to the receptor, as overexpression or mutations in this protein are associated with tumor development.

The ACK2 antibody is widely utilized as a marker to identify hematopoietic progenitors, and to neutralize receptor-ligand binding in vitro and in vivo (use format appropriate for functional assays). In addition, the antibody is reported to be cross-reactive with rat c-Kit and is extensively published for use in this species.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: ACK2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty