Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD4 Mouse Monoclonal Antibody (violetFluor® 450) [clone: RPA-T4]

Supplier: Tonbo Biosciences
75-0049-T025 75-0049-T100
10051-774EA 159 USD InStock
10051-774 10051-772
Anti-CD4 Mouse Monoclonal Antibody (violetFluor® 450) [clone: RPA-T4]
Antibodies
The RPA-T4 antibody reacts with human CD4, a 59 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.

The RPA-T4 antibody is widely used as a phenotypic marker for human CD4 expression, and is cross-reactive with Chimpanzee CD4. This antibody recognizes a different epitope, and thus does not block binding, of the alternative Anti-Human CD4 antibody clone OKT4 (Reinherz EL, et al. 1979. Proc. Natl. Acad. Sci. 76:4061-4065).

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty