Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Foxp3 / Transcription Factor Staining Buffer Kit, Tonbo Biosciences

Supplier: Tonbo Biosciences
TNB-1022-L160 TNB-0607-KIT TNB-1213-L150 TNB-1020-L050
10051-874EA 143 USD InStock
10051-874 10051-876 10051-878 10141-138
Foxp3 / Transcription Factor Staining Buffer Kit, Tonbo Biosciences
Assays Protein Assays
Tonbo Biosciences Foxp3 / Transcription Factor Staining Buffer Kit contains specially formulated buffers and solutions for optimal resolution and low background in your analysis of nuclear antigens by flow cytometry

Foxp3 / Transcription Factor Fix/Perm Concentrate (4X) 50 mL. A concentrated solution which, when diluted with Foxp3 / Transcription Factor Fix/Perm Diluent (1X) provides best results in protocols for intranuclear staining of transcription factors using fluorescently conjugated antibodies.

Foxp3 / Transcription Factor Fix/Perm Diluent (1X) 160 mL. Intended for use as a diluent for Foxp3 / Transcription Factor Fix/Perm Concentrate (4X).

Flow Cytometry Perm Buffer (10X) 150 mL. Provided as a concentrate which, when diluted with distilled water to a 1X solution, provides best results in intracellular staining protocols for cytokines and other cytoplasmic antigens, by maintaining membrane permeabilization throughout staining and wash steps.

This complete kit contains three components for use in these staining protocols for detection of nuclear antigens such as Foxp3 and ROR gamma.

Tested applications: Flow Cytometry
Reactivity: Mouse, Human
Storage temperature: Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.

Ordering information: Kit includes (1) Foxp3 / Transcription Factor Fix/Perm Concentrate (4X) 50 mL. A concentrated solution which, when diluted with Foxp3 / Transcription Factor Fix/Perm Diluent (1X) provides best results in protocols for intranuclear staining of transcription factors using fluorescently conjugated antibodies; (2) Foxp3 / Transcription Factor Fix/Perm Diluent (1X) 160 mL. Intended for use as a diluent for Foxp3 / Transcription Factor Fix/Perm Concentrate (4X); (3) Flow Cytometry Perm Buffer (10X) 150 mL. Provided as a concentrate which, when diluted with distilled water to a 1X solution provides best results in intracellular staining protocols for cytokines and other cytoplasmic antigens, by maintaining membrane permeabilization throughout staining and wash steps.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty