Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CREB Mouse Monoclonal Antibody [clone: 10E9]

Supplier: Enzo Life Sciences
905-639-100
10051-898EA 556 USD InStock
10051-898
Anti-CREB Mouse Monoclonal Antibody [clone: 10E9]
Antibodies
The transcription factor CREB (cAMP response element binding) binds the palindromic octanucleotide TGACGTCA. Members of the ATF/CREB family include CREB-1, CREB-2 (also designated ATF-4), ATF-1, ATF-2, and ATF-3. Phosphorylation of CREB allows recruitment of CREB-binding protein (CBP), a large co-activator that contacts the general transcriptional machinery. CREB-mediated transcription regulates diverse cellular responses, including intermediary metabolism, neuronal signaling, cell proliferation, and apoptosis.

Type: Primary
Antigen: CREB
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty