Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DopamineR D2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-740-100
10051-934EA 452 USD InStock
10051-934
Anti-DopamineR D2 Rabbit Polyclonal Antibody
Antibodies
Dopamine is the predominant catecholamine neurotransmitter in the mammalian brain, where it regulates locomotor, emotional, memory, and dietary intake functions, as well as peripheral roles such as hormone secretion and renal function. Dopamine receptors (D1-D5) are members of the seven transmembrane domain containing GPCR family, and are the target of several anti-psychosis and anti-Parkinsonian drugs. The D2, D3, and D4 receptors signal via coupling of Gi/Go alpha G-protein subunits to inhibit adenylyl cyclase activity and modulate cellular potassium and calcium ion channel conductance. Expression of the D2 receptor is found primarily in the brain (striatum, olfactory tubercle, and nucleus accumbens) and in the anterior and intermediate lobes of the pituitary.

Type: Primary
Antigen: DopamineR D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-Terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty