Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR7 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-CSA-824-E
10051-980EA 539 USD InStock
10051-980
Anti-TLR7 Rabbit Polyclonal Antibody
Antibodies
The TLRs (Toll-like receptors) are a group of evolutionarily conserved pattern recognition molecules that play a key role in host defense during microbial infection by regulating both innate and adaptive immune responses. TLRs are type I transmembrane proteins with ectodomains containing interspersed leucine-rich repeat (LRR) motifs that are involved in the recognition of various microbial ligands.

Type: Primary
Antigen: TLR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty