Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Hemostat Blood, Rabbit, Sterile, Hardy Diagnostics
Supplier: Hardy Diagnostics
RB250 RB050 RB100
10052-820EA 107.13 USD InStock
10052-820 10052-824 10052-822
Hemostat Blood, Rabbit, Sterile, Hardy Diagnostics
Blood
Blood, rabbit, sterile, screw cap
Our animal blood is obtained from a controlled herd that is kept free of antimicrobic drugs and disease
All types of animal blood and packaging configurations are available.
All animals are maintained in a healthy condition and treated humanely in accordance with government regulations. Fresh blood is available every week.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty