Would you like to log in again?
Fletcher's Medium Broth Base
Supplier: Hardy Diagnostics
SRS125
10052-852EA 191.56 USD InStock
10052-852
Fletcher's Medium Broth Base
Media for Microbiology Dehydrated Media for Microbiology
For the culture of Leptospira.
Ordering information: Supplement required, rabbit serum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty