Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GOLD BRAND ASTM/API High Precision Plain Form Hydrometer, Thermco

Supplier: Thermco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ACC025H ACC039H ACC027H ACC040H ACC023H ACC024H ACC033H ACC031H ACC035H ACC001H ACC010H ACC021H ACC008H ACC022H ACC037H ACC036H ACC034H ACC002H ACC026H ACC032H ACC038H ACC009H ACC004H ACC007H ACC005H ACC028H ACC029H ACC003H ACC030H ACC006H
10053-098EA 65.14 USD InStock
10053-098 10053-076 10053-054 10053-078 10053-056 10053-058 10053-090 10053-070 10053-092 10053-072 10053-094 10053-050 10053-074 10053-096 10053-052 10053-086 10053-064 10053-100 10053-066 10053-088 10053-102 10053-046 10053-068 10053-104 10053-048 10053-080 10053-060 10053-082 10053-062 10053-084
GOLD BRAND ASTM/API High Precision Plain Form Hydrometer, Thermco
Hydrometers
High precison ASTM plain form hydrometers made to exceed ASTM and API specifications, which do not incorporate a thermometer in the hydrometer

The temperature standardization is 60/60F. The hydrometer scale division are 0.1 degree.

Tall form, 330 mm, has 12 degree ranges; Short form, 163 mm, has 6 degree ranges.

Used to measure the density according to API degree scale for petroleum applications.

Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.

Ordering information: Hard square plastic tube included.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty