Would you like to log in again?
ASTM/API Precision Combined Form +30/180 ºF Hydrometer, Thermco
Supplier: Thermco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GW052HMS GW060HMS GW054HMS GW057HMS GW051HMS GW055HMS GW059HMS GW043HMS GW041HMS GW058HMS GW056HMS GW053HMS GW042HMS GW045HMS GW044HMS
10053-276EA 82.49 USD InStock
10053-276 10053-540 10053-550 10053-278 10053-542 10053-552 10053-544 10053-554 10053-532 10053-546 10053-556 10053-534 10053-548 10053-536 10053-538
ASTM/API Precision Combined Form +30/180 ºF Hydrometer, Thermco
Hydrometers
Precison ASTM-like combined form hydrometers made to meet ASTM and API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 30 to 180 degrees farenheit and is mounted in the body of the hydrometer. Length of hydrometer is 380mm.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty