Would you like to log in again?
API Precision Short Plain Form Hydrometer, Thermco
Supplier: Thermco
GW2505 GW2506 GW2504 GW2508 GW2503 GW2507 GW2502 GW2500 GW2509 GW2501
10053-296EA 45.68 USD InStock
10053-296 10053-284 10053-298 10053-286 10053-288 10053-290 10053-292 10053-280 10053-294 10053-282
API Precision Short Plain Form Hydrometer, Thermco
Hydrometers
Precison API plain form hydrometers made to meet API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers.
Length of the hydrometer is 6 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty