Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ASTM/API Precision Plain Form Hydrometer, Thermco

Supplier: Thermco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

GW030H GW037H GW006H GW021H GW036H GW002H GW004H GW010H GW034H GW035H GW029H GW031H GW023H GW003H GW033H GW040H GW025H GW032H GW008H GW007H GW039H GW009H GW028H GW024H GW038H GW022H GW026H GW001H GW005H
10053-494EA 41.12 USD InStock
10053-494 10053-274 10053-152 10053-174 10053-496 10053-176 10053-154 10053-498 10053-156 10053-158 10053-490 10053-170 10053-492 10053-272 10053-150 10053-172 10053-164 10053-186 10053-166 10053-484 10053-168 10053-486 10053-500 10053-488 10053-180 10053-182 10053-160 10053-184 10053-162
ASTM/API Precision Plain Form Hydrometer, Thermco
Hydrometers
Precison ASTM plain form hydrometers made to meet ASTM and API specifications, which do not incorporate a thermometer in the hydrometer

The temperature standardization is 60/60F. The hydrometer scale division are 0.1 degrees.

Tall form, 330mm, have 12 degree ranges; Short form, 163mm, have 6 degree ranges.

Used to measure the density according to API degree scale for petroleum applications.

Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty