Would you like to log in again?
ASTM/API Precision Combined Form 0/150 ºF Hydrometer, Thermco
Supplier: Thermco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GW056HLS GW054HLS GW045HLS GW053HLS GW059HLS GW058HLS GW051HLS GW057HLS GW044HLS GW042HLS GW055HLS GW052HLS GW043HLS GW041HLS GW060HLS
10053-520EA 83.58 USD InStock
10053-520 10053-530 10053-522 10053-510 10053-524 10053-502 10053-512 10053-526 10053-504 10053-514 10053-528 10053-506 10053-516 10053-508 10053-518
ASTM/API Precision Combined Form 0/150 ºF Hydrometer, Thermco
Hydrometers
Precison ASTM-like combined form hydrometers made to meet ASTM and API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and is mounted in the body of the hydrometer. Length of hydrometer is 380mm.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty