Would you like to log in again?
API Precision Combined Form Hydrometer, Thermco
Supplier: Thermco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GW6004S GW6006S GW6002S GW6005S GW6009S GW6007XS GW6001XS GW6007S GW6008S GW6001S GW6000S GW6000XS GW6003S
10053-582EA 82.49 USD InStock
10053-582 10053-592 10053-570 10053-584 10053-594 10053-572 10053-586 10053-574 10053-588 10053-576 10053-578 10053-580 10053-590
API Precision Combined Form Hydrometer, Thermco
Hydrometers
Precison API combined form hydrometers made to meet API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and mounted in the body of the hydrometer. Length of the hydrometer is 15 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty