Would you like to log in again?
Personna® 2-Facet Aluminum Back Single Edge Blades and Dispenser, AccuTec Blades
Supplier: AccuTec Blades
66-0210-0000
10053-717CS 378.25 USD InStock
10053-717
Personna® 2-Facet Aluminum Back Single Edge Blades and Dispenser, AccuTec Blades
Razors and Blades
The Personna® 0.009" single edge razor blade dispensers are the safest way to handle and store blades
All blades in the dispenser are manufactured from high-quality carbon knife steel.
The clear plastic dispenser contains 10 blades and allows for easy viewing of unused blades. Blades easily slide out of the dispenser with no shelling paper to remove. Used blades slide into the bottom of the dispenser for storage until disposal.
Ordering information: All blades are supplied 10 per dispenser. Each pack contains 6 dispensers and each case contains 72 dispensers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty